Learn More
Essendant GLASSESREADR BK/CLR 1.5
Supplier:  Essendant ANS19878
Encompass_Preferred
Safety eyewear readers have a stylish and ergonomic nylon frame High-performance polycarbonate lenses are sturdy and provide a clear field of vision Precise placement of diopter insert within lens helps prevent eyestrain headaches and accidents Soft comfortable rubber nose pad is self-adjusting and fits well on almost any wearer Lenses provide 99 9pct UV protection Eyewear meets ANSI Z87 1plus impact standard plus 1 5 Diopter Strength Safety Glasses Style- Wraparound Frame Color- Black Frame Material- Nylon Lens Material- Polycarbonate
Catalog No. 50-000-22992