Learn More
Essendant GLASSESSAFETYANTI-SCLR
Supplier:  Essendant MMM1122008000
Encompass_Preferred
Economy safety glasses that combine style protection and affordability to safeguard workers on jobsites Providing a wide viewing area the wraparound design shields to keep eyes more fully protected Contoured temples fit comfortably and snugly without pressure points but also without any looseness that causes shifting Recommended for use during chipping chiseling drilling grinding machining masonry pouring/casting power fastening riveting sanding and sawing Safety Glasses Style- Wraparound Frame Color- Clear Frame Material- Polycarbonate Lens Material- Polycarbonate
Catalog No. 50-000-34013