Learn More
Essendant GLASSESSAFETYCLR
Supplier:  Essendant MMM118190000020
Encompass_Preferred
An economy safety glass that offers style protection and affordability Providing a wide viewing area the wraparound design combines with the integrated side shields to keep eyes more fully protected Contoured temples fit comfortably and snugly without pressure points but also without any looseness that causes shifting The impact-resistant polycarbonate lens absorbs 99 9pct UV rays Recommended for use during chipping chiseling drilling grinding machining masonry pouring/casting power fastening riveting sanding and sawing Safety Glasses Style- Frameless Frame Color- Clear Frame Material- Polycarbonate Lens Material- Polycarbonate
Catalog No. 50-000-33416