Learn More
Essendant GLASSESSAFETYUVBK
Supplier:  Essendant ANS21917
Encompass_Preferred
Stylish safety eyewear is designed to fit over most prescription eyeglasses Integral side shields offer increased protection Lenses offer 99pct UV protection Eyewear meets ANSI Z87 1plus and CSA Z94 3 standards Safety Glasses Style- Wraparound Frame Color- Black Lens Coating/Filter- IR/UV 5 0
Catalog No. 50-000-22805