Learn More
Essendant GLASSESSFTYCLR LEN
Supplier:  Essendant ANS22608
Encompass_Preferred
KleenGuard Personal Protective Equipment (PPE) enables rather than inhibits allowing for maximum productivity and safety for you and your employees Giving your employees what they need to do their job safely and effectively is a top priority of any business owner KleenGuard Nemesis Safety Eyewear s stylish good looks and durable lightweight construction are just some of the reasons why millions of people wear them for protection both at work and at home Nemesis Safety Glasses have KleenVision Anti-Fog coated polycarbonate lenses that provide 99 9pct UVA/UVB/UVC protection to help prevent eye damage like cataracts retinal damage and other conditions that can cause temporary vision loss The wraparound frame has a base curve of eight that extends the range of peripheral protection as well as soft-touch temples for comfort The Nemesis safety glasses have a patented nose piece designed to be extremely flexible to channel sweat away from the eyes and to help prevent slippage down
Catalog No. 50-000-22873