Learn More
Essendant GLASSESSFTYDUAL RDRS6
Supplier:  Essendant ANS20387
Encompass_Preferred
KleenGuard Personal Protective Equipment (PPE) enables rather than inhibits allowing for maximum productivity and safety for you and your employees When you need vision assistance and eye protection you can trust KleenGuard Dual Readers They feature integrated diopters on both the top and the bottom for vision help when looking both up and down The stylish nylon frames feature a soft self-adjusting rubber nose pad for comfort The lenses are hard coated for scratch resistance and longer life These safety glasses meet ANSI Z87 1plus standards for high impact and the polycarbonate lenses provide 99 9pct UVA/UVB/UVC protection Making the right Personal Protection choice is difficult Perform with distinction with KleenGuard Personal Protective Equipment (PPE) Safety Glasses Style- Wraparound Frame Color- Black Frame Material- Polycarbonate Lens Material- Polycarbonate
Catalog No. 50-000-22825