Learn More
Electron Microscopy Sciences Glassine Negative Envelope Plain, 7 x 7 cm
SDP
Supplier:  Electron Microscopy Sciences 74087
Encompass_Preferred
Glassine envelopes, acid free for negative storage. Open end, thumb-cut, side-seamed.
Use: 7 x 7 cm
Catalog No. 50-261-63