Learn More
Cayman Chemical GlatIrameracetate 5mg
SDP
Supplier:  Cayman Chemical 90005715 MG
Encompass_Preferred
A mixture of synthetic polypeptides that prevents MBP to MCH class II molecules; increases oligodendrocyte and neuronal precursor cell proliferation
Catalog No. 50-289-2427