Learn More
Medchemexpress LLC HY-N2112 5mg Medchemexpress, Glaucocalyxin A CAS:79498-31-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN21125MG
Encompass_Preferred
Medchemexpress, HY-N2112 5mg Glaucocalyxin A CAS:79498-31-0 Glaucocalyxin A, an ent-kauranoid diterpene from Rabdosia japonica var., induces apoptosis in osteosarcoma by inhibiting nuclear translocation of Five-zinc finger Glis 1 (GLI1) via regulating PI3K/Akt signaling pathway. Glaucocalyxin A has antitumor effect. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-1481