Learn More
Medchemexpress LLC GLB1/Beta-galactosidase antibody (YA2816) | 50 UL

Supplier: Medchemexpress LLC HYP8307150UL
GLB1/Beta-galactosidase Antibody (YA2816) is a rabbit-derived, non-conjugated IgG monoclonal antibody targeting GLB1/Beta-galactosidase. It is developed against the glycosyl hydrolase 35 family of proteins, which catalyzes the hydrolysis of a terminal beta-linked galactose residue. Mutations in the related gene may result in GM1-gangliosidosis and Morquio B syndrome.
- Host: Rabbit
- Reactivity: Human, mouse, rat
- Conjugation: Non-conjugated
- Purity: Affinity purified
- Appearance: Liquid
- Applications: WB, FC
- Formulation: Supplied in 10mM phosphate buffered saline, pH 7.4, 150mM sodium chloride, 0.05% BSA, 0.02% sodium azide and 50% glycerol.
- Storage conditions: Stored at -20°C for 1 year. Avoid repeated freeze/thaw cycles.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.