Learn More
Medchemexpress LLC Glecaprevir | 1365970-03-1 | 838.87 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY17634100MG
Encompass_Preferred
Glecaprevir is a novel HCV NS3/4A protease inhibitor and a SARS-CoV 3CLpro inhibitor. It is intended for use as a laboratory chemical and for the manufacture of substances, exclusively for research purposes and by experienced personnel.
  • HCV NS3/4A protease inhibitor
  • SARS-CoV 3CLpro inhibitor
  • IC50 values ranging from 3.5 to 11.3 nM (HCV NS3/4A protease)
  • IC50 of 4.09 μM (SARS-CoV 3CLpro)
  • For research use only
  • Intended for experienced personnel
Catalog No. 50-003-86988