Learn More
Selleck Chemical LLC Glecaprevir 1mg 1365970-03-1
SDP
Supplier:  Selleck Chemical LLC S57201MG
Encompass_Preferred
Glecaprevir is a hepatitis C virus (HCV) nonstructural (NS) protein 3/4A protease inhibitor that targets the the viral RNA replication. It displays IC50 values ranging from 3.5 to 11.3 nM for clinical isolates of HCV genotypes 1a, 1b, 2a, 2b, 3a, 4a, 5a, and 6a. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8413