Learn More
AUTOMATION TRAINER GLH QC Kit - DMSO Reagents, 96-Well Verification Microplates, VRP, Mini-Reader
SDP
Supplier:  AUTOMATION TRAINER ATQCKITDMSO96
Encompass_Preferred
Quality Simplified. The vision behind the GLH QC Kit was to provide users with a simplified, accurate, and cost-effective quality control solution.Test your automated liquid handlers, multi-channel pipettes, readers, labware, and more. Develop unique liquid classes and Optimize Assays.- Characterizes error limits of volume transfers (1 nL to 10 mL)- Compatible with any manual or automated liquid handling instrument (up to 1536-channels)- Test Serial Dilutions (up to 48 targets in a single plate)- Diluent-Dispense-Insensitive with GLH Verification Microplates- No manual pipetting required with 96-Well and 384-Well GLH Verification Microplates- Correct any reader for NIST-Traceable results with the GLH Validation Reference PlateLegal Disclaimer: For research use only, not for diagnostic procedures.
Catalog No. NC2293252
Request a Quote
Please contact your sales representative for a quote or further ordering instructions.