Learn More
AUTOMATION TRAINER GLH QC Kit Lite - DMSO Reagents Set and 96-Well Verification Microplates
SDP
Supplier:  AUTOMATION TRAINER ATQCKITLTDMSO96
Encompass_Preferred
The vision behind the GLH QC Kit was to provide users with a simplified, accurate, and cost-effective quality control solution. Test your automated liquid handlers, multi-channel pipettes, readers, labware, and more. Develop unique liquid classes and Optimize Assays.Characterizes accuracy and precision of volume transfersCan be used for hardware calibration of automated liquid handlers and manual pipettors- Two sets of reagents- GLH-Tracker Desktop Software- MicroplatesLegal Disclaimer: For research use only, not for diagnostic procedures.
Catalog No. NC2293257
Please contact your sales representative for a quote or further ordering instructions.