Learn More
Medchemexpress LLC Glimepiride | 1131981-29-7 | 99.4% | C24H30D4N4O5S | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB0104S11MG
Encompass_Preferred
Glimepiride-d4-1 is a deuterium-labeled version of Glimepiride, a medium-to-long acting sulfonylurea anti-diabetic compound. This labeled compound can be used for quantitative analysis and tracing in various scientific applications.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuteration may influence pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-32823