Learn More
Medchemexpress LLC Glimepiride | 93479-97-1 | 99.9% | C24H34N4O5S | 10 MM * 1 ML
SDP
Supplier:  Medchemexpress LLC HYB010410MM/1ML
Encompass_Preferred
Glimepiride is a medium-to-long acting sulfonylurea anti-diabetic compound with an ED50 of 182 μg/kg. It is for research use only. It has been shown to decrease extracellular Aβ40 and Aβ42 levels, potentially serving as a promising drug for the treatment of AD associated with diabetes. Compared to other sulfonylureas, it is generally associated with a lower risk of hypoglycemia and less weight gain, and may be safer for patients with cardiovascular disease due to its lack of detrimental effects on ischemic preconditioning.
  • Medium-to-long acting sulfonylurea anti-diabetic compound
  • Lowers blood glucose in rabbits more effectively than glibenclamide
  • Associated with lower risk of hypoglycemia and less weight gain
  • May be safer for patients with cardiovascular disease
Catalog No. 50-004-32714