Learn More
Medchemexpress LLC Glimepiride sulfonamide | 119018-29-0 | MFCD02955393 | 98.0% | 351.43 g/mol | C16H21N3O4S | 50 MG

Supplier: Medchemexpress LLC HYW04132450MG
Glimepiride sulfonamide is a chemical intermediate used as a precursor in the synthesis of the antidiabetic agent glimepiride. It is supplied as a solid and as DMSO solutions for analytical or synthetic use, and is intended for research use only (not for human use).
- Chemical intermediate for synthesis of glimepiride.
- Molecular formula C16H21N3O4S.
- Molecular weight 351.43 g/mol.
- Typical purity 98.0%.
- Available as solid (50 MG, 100 MG, larger sizes) and 10 mM solutions in DMSO.
- For research use only; not for clinical or human use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.