Learn More
Selleck Chemical LLC Gliocidin E5804-25MG
SDP
Supplier:  Selleck Chemical LLC E580425MG
Encompass_Preferred
Gliocidin (SW106065) is a nicotinamide-mimetic prodrug that targets glioblastoma cells with an IC50 of 150 nM by exploiting a de novo purine synthesis vulnerability via indirect inhibition of IMPDH2 It readily crosses the BBB and can be used in the treatment of all malignant brain tumors
Catalog No. 50-000-40876