Learn More
Medchemexpress LLC HY-B0254 100mg Medchemexpress, Glipizide CAS:29094-61-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0254100MG
Encompass_Preferred
Medchemexpress, HY-B0254 100mg Glipizide CAS:29094-61-9 Glipizide (CP 2872; K 4024) a potent, orally active and sulfonylurea class anti-diabetic agent and can be used for type 2 diabetes mellitus research but not type 1. Glipizide acts by partially blocking ATP-sensitive potassium (KATP) channels among β cells of pancreatic islets of Langerhans. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2427