Learn More
Apexbio Technology LLC Glipizide 29094-61-9 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC A84385.1
Encompass_Preferred
Glipizide (CAS 29094-61-9) is a small molecule that acts by inhibiting ATP-sensitive potassium channels in pancreatic cells leading to membrane depolarization and subsequent stimulation of insulin secretion Additionally it modulates ATP-dependent K channels in GABAergic neurons within the brain Glipizide is widely used in research to investigate pancreatic cell physiology insulin release mechanisms and the role of KATP channels in neuroendocrine regulation
Catalog No. 50-000-96238