Learn More
Medchemexpress LLC Glipizide (Standard) | 29094-61-9 | MFCD00870381 | 99.7% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB0254R50MG
Encompass_Preferred
Glipizide (Standard) is the analytical standard of Glipizide. It is intended for research and analytical applications. Glipizide (CP 2872; K 4024) is a potent, orally active sulfonylurea class anti-diabetic agent used for type 2 diabetes mellitus research, but not for type 1. It functions by partially blocking ATP-sensitive potassium (KATP) channels in the β cells of pancreatic islets of Langerhans.
  • Supplied as an assay reference standard.
  • Commonly used in qualitative, quantitative, and methodological research experiments, including HPLC, GC, and MS.
  • Potent, orally active sulfonylurea class anti-diabetic agent.
  • Partially blocks ATP-sensitive potassium (KATP) channels in pancreatic β cells.
Catalog No. 50-004-06963