Learn More
Aviva Systems Biology Corp GLIS3 Antibody - N-terminal region (ARP39924_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39924_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against GLIS3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human GLIS3. Alternate names: ZNF515ApplicationIHC, WB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53161