Learn More
ABclonal Technology GLMN Rabbit pAb
SDP
Supplier:  ABclonal Technology A1202520UL
Encompass_Preferred
This gene encodes a phosphorylated protein that is a member of a Skp1-Cullin-F-box-like complex. The protein is essential for normal development of the vasculature and mutations in this gene have been associated with glomuvenous malformations, also called glomangiomas. Multiple splice variants encoding different isoforms have been found for this gene.
Catalog No. 50-001-14138