Learn More
Medchemexpress LLC HY-101710 20mg Medchemexpress, GLN-1062 CAS:224169-27-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10171020MG
Encompass_Preferred
Medchemexpress, HY-101710 20mg GLN-1062 CAS:224169-27-1 Memogain is a pro-drug of galantamine, used for the treatment for Alzheimer’s disease. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2197