Learn More
Cayman Chemical Globosuxnthon A 5mg
SDP
Supplier:  Cayman Chemical 274865 MG
Encompass_Preferred
A fungal metabolite; cytotoxic to various human solid tumor cell lines, including MCF-7, PC3, LNCap, and DU145 cells (IC50s = 1.3, 0.65, 1.5, and 1.2 µM, respectively); active against C. albicans
Catalog No. 50-294-0090