Learn More
STEMCELL Technologies GloCell™ Fixable Viability Dye Violet 540, Size: 500 Tests
SDP
Supplier:  STEMCELL Technologies 75011
Encompass
GloCell™ Fixable Viability Dye Violet 540 is a fluorescent cell viability dye for staining live/dead mammalian cells in applications such as flow cytometry. The dye irreversibly binds intracellular and cell surface amine groups and can be utilized prior to fixation, permeabilization, or cryopreservation. Cells with compromised plasma membranes become permeable to the GloCell™ dye, resulting in a greater fluorescence compared to live cells in a cell viability assay.GloCell™ Fixable Viability Dye Violet 540 is excited by the violet laser at 405 nm and it has a fluorescence emission maximum of 540 nm that can be detected using the 525/50 band pass filter used to detect AmCyan.
Catalog No. 50-197-6221