Learn More
STEMCELL Technologies GloCell™ Fixable Viability Dye Red 710, Size: 500 Tests
SDP
Supplier:  STEMCELL Technologies 75006
Encompass
GloCell™ Fixable Viability Dye Red 710 is a fluorescent cell viability dye for staining live/dead mammalian cells in applications such as flow cytometry. The dye irreversibly binds intracellular and cell surface amine groups and can be used prior to fixation, permeabilization, or cryopreservation. Cells with compromised plasma membranes become permeable to the GloCell™ dye, resulting in greater fluorescence compared to live cells in a cell viability assay.GloCell™ Fixable Viability Dye Red 710 is excited by the red laser (633 nm) with an excitation maximum of 647 nm, and it has a fluorescence emission maximum of 710 nm that can be detected using the 710/50 band pass filter used to detect Alexa Fluor 700.
Catalog No. 50-197-6211