Learn More
Med Vet International Exam Glove STERLING Medium NonSterile Nitrile Standard Cuff Length Textured Fingertips Gray Chemo Tested, 50707, 200/Box
Supplier:  Med Vet International 747112BX
Encompass_Preferred
Nitrile material allows for comfortable wear Textured fingertips provide reliable grip Beaded cuffs help keep the gloves in place Chemo tested (Cyclophosphamide, Doxorubicin HCl, Etoposide, 5-Fluorouracil, Paclitaxel (Taxol), Cisplatin, Decarbonize, Ifosfamide, Mitoxantrone, Vincristine Sulfate) Nonsterile and designed for one-time use Like all medical gloves, these are powder free Medium
Catalog No. 50-304-4301