Learn More
Med Vet International Exam Glove Halyard™ Lavender™ Medium NonSterile Nitrile Standard Cuff Length Textured Fingertips Lavender Not Rated, 52818, 250/Box
Supplier:  Med Vet International 678086BX
Encompass_Preferred
Lightweight and comfortable Halyard Lavender nitrile gloves are suitable for tasks such as oral care, physical exams and dispensing of medications Flexible material allows for full dexterity and a free range of finger movement A powder- and latex-free design reduces reactions in sensitive skin The ambidextrous design fits either hand Textured fingertips provide a sure grip on containers and implements These Halyard exam gloves come in a Smartpull dispenser box with two openings
Catalog No. 50-304-3675