Learn More
Med Vet International McKesson Confiderm LDC Nitrile Exam Glove, Small, Blue, 14-6N42C, 250/Box
Supplier:  Med Vet International 1159329BX
Encompass_Preferred
Exam Glove McKesson Confiderm LDC Small NonSterile Nitrile Standard Cuff Length Fully Textured Blue Chemo Tested / Fentanyl Tested Low dermatitis potential; ideal for individuals with skin sensitivities to regular nitrile gloves Tested to resist permeation of 15 chemotherapy drugs, plus fentanyl Exterior's fully textured finish promotes slip resistance in wet or dry conditions Thin with high-tactile sensitivity Excellent conformability provides superior fit and extended user comfort Nitrile material is durable with tensile strength Fingertip thickness: 0.07mm (2.8 mil) Chlorinated internal surface Like all medical gloves, these are powder free Small
Catalog No. 50-304-4144