Learn More
Med Vet International Exam Glove Trilon X-Large NonSterile Vinyl Standard Cuff Length Smooth Clear Not Rated, 25-98, 100/Box
Supplier:  Med Vet International 287772BX
Encompass_Preferred
A high tensile strength makes these medical exam gloves sturdy Supple and soft construction allow for a full range of movement These Trilon gloves are ambidextrous The tear-resistant, puncture-resistant and abrasion-resistant gloves are durable and long-lasting The vinyl, powder-free exam gloves contain no latex to help ensure comfort
Catalog No. 50-304-4995