Learn More
Med Vet International Dynarex Powder-Free Plus Latex Exam Gloves, Polylined, Small, 1000/Case
Supplier:  Med Vet International 6612
Encompass_Preferred
Dynarex Powder-Free Plus Latex Exam Gloves are crafted from a specially formulated, non-chlorinated latex material for optimal performance and durability. The ambidextrous gloves are designed with an embossed micro-textured surface and interior for increased grip. The gloves feature a minimum thickness of 5 mil at the palm and 6 mil at the fingertips for optimal puncture resistance. A polymer coating and beaded cuff ensure easy donning and prevent tears. The gloves fit securely around the wrist and have h and -conforming flexibility to prevent physical fatigue. Frequently used in many settings; including veterinary clinics, retirement homes, and medical facilities. Dynarex Powder-Free Plus Latex Exam Gloves offer superior comfort for sensitive h and s. The gloves have a micro-textured surface that allows for increased grip during delicate procedures. A minimum thickness of 5 mil at the palm and 6 mil at the fingertips provides added protection and puncture resistance.
Catalog No. 50-283-0020