Learn More
Med Vet International SW TrueForm TF-95RB Royal Blue Large Powder Free Disposable Gloves - Medical Exam Grade - 9.5 in Length - Textured Fingers Finish - 3.1 mil Thick - N301344, 100/Box
Supplier:  Med Vet International N301344
Encompass_Preferred
Product SpecificationsBrand: SW SafetyProduct Type: Disposable GlovesGrade: Medical ExamMaterial: NitrilePowdering: Powder FreeThickness: 3.1 milOverall Length: 9.5 inColor: Royal BlueSize: LargeSurface Finish: Textured FingersResistant Properties: Chemical-ResistantCuff Type: BeadedSeries: TF-95RBStandards Met: SHA Dermatologically ApprovedPackage Quantity: 100 per box, 10 boxes per caseTrade Name: TrueForm
Catalog No. 50-252-2231