Learn More
World Precision Instrument Glover Bulldog Clamp,Straight, smooth tips
SDP
Supplier:  World Precision Instrument 503238
Encompass_Preferred
  • Straight, smooth tips
  • 7cm long
  • Tension adjustment
Catalog No. 50-252-1277