Learn More
Essendant Safety Zone ProGuard Powder Free Nitrile Gloves, Powder-Free, X-Large, 3.5 mil Thick, 9.65" Long, Black, 100/Box, 10 Boxes/Carton
Supplier:  Essendant SZNGNPRXLBK
Encompass_Preferred
Nitrile gloves are the top choice for professionals who demand superior quality and reliability With exceptional strength and durability these gloves are perfect for tasks that require protection against punctures cuts and abrasions They offer excellent barrier protection against chemicals oils and other hazardous materials making them ideal for environments where workers are exposed to harmful substances Nitrile gloves are also designed to fit snugly on the hand providing a comfortable and flexible feel that allows for ease of movement and tactile sensitivity Latex-free and hypoallergenic Glove Material- Nitrile Packaged Sterile- No Size Type- X-Large Length- 9 65
Catalog No. 50-000-30047