Learn More
Essendant GLOVES4M L NITRSHD PWD F
Supplier:  Essendant MPG6015L
Encompass_Preferred
Resistance to a wide variety of chemicals Latex free Glove Material- Nitrile Packaged Sterile- No Size Type- Large Thickness- 4 mil
Catalog No. 50-000-31225