Learn More
Essendant GLOVESCUT RSTL12PRGY
Supplier:  Essendant ANS13825
Encompass_Preferred
KleenGuard Personal Protective Equipment (PPE) enables rather than inhibits allowing for maximum productivity and safety for you and your employees KleenGuard G60 Level 3 Polyurethane Coated Cut Resistant Gloves help give your team a confident level of protection against hand injuries and lacerations They are ANSI Cut Level 3 (CPPT 1 010 grams) and EN388 Cut Level 3 protective gloves These coated safety gloves are made of ultra-high molecular weight polyethylene (UHMWPE) fiber which enhances breathability and protection for a comfortable fit The polyurethane coated palm and fingers give your workers excellent grip and dexterity so they won t sacrifice productivity for protection Each pair has a dedicated right and left glove for a snug fit Making the right Personal Protection choice is difficult Perform with distinction with KleenGuard Personal Protective Equipment (PPE) ANSI Cut Resistance- A3 Anti-Vibration- No EN 388 Badge- 4X42B Size Type- Large
Catalog No. 50-000-20403