Learn More
Essendant GLOVESCUTPRO 92721XXLBK
Supplier:  Essendant CRW92721XXL
Encompass_Preferred
MCR Safety s Cut Pro Series offers various cut protection levels and materials for your protection The 92721 cut-resistant work glove is made with a 21-gauge HyperMax shell and features extreme dexterity It also provides good abrasion and puncture resistance making it the perfect glove for demanding jobs The 92721 has a black polyurethane palm and fingertip coating that easily hides dirt and grime ANSI Cut Resistance- A4 ANSI Abrasion Resistance- Level 3 ANSI Puncture Resistance- Level 2 Anti-Vibration- No
Catalog No. 50-000-19767