Learn More
Essendant GLOVESDIPDZMBEBK
Supplier:  Essendant CRWN9696M
Encompass_Preferred
Gloves for protection comfortable fit and sure grip Resistant to cuts and abrasions Provides tactile sensitivity and hand dexterity ANSI Cut Resistance- A1 ANSI Abrasion Resistance- Level 3 ANSI Puncture Resistance- Level 2 Anti-Vibration- No
Catalog No. 50-000-18832