Learn More
Essendant GLOVESFOAMSZ 812PR/PK
Supplier:  Essendant ANS40226
Encompass_Preferred
These gloves perform in a wide range of applications providing outstanding grip dexterity and protection against minor cuts punctures and abrasions Even enables you to maintain a steady grip when working around oil and dirt Washable-maintains the EN388 ratings through at least five wash cycles Rated for EN1149-2 Vertical Resistance Improved heat transfer stamp-ratings are visible even after glove use Anti-Vibration- No EN 388 Badge- 3121X Size Type- Medium Size Number- Size 8
Catalog No. 50-000-22916