Learn More
Essendant GLOVESGPNITRILEPFMBE
Supplier:  Essendant BWK395MBXA
Encompass_Preferred
For superior protection against chemicals and sterile handling of food these latex-free disposable gloves combine strength and convenience of single-use protective handwear The material is chemical-resistant for safe handling of dangerous liquids The strong design is puncture-resistant for added protection against sharp objects and abrasions Ideal for lab work chemical handling and food service applications where safety and cleanliness are priorities 395 Series Glove Material- Nitrile Packaged Sterile- No Size Type- Medium Length- 9 5
Catalog No. 50-000-21251