Learn More
Essendant GLOVESHPPENITRILEXLBK
Supplier:  Essendant CRW92720NFXL
Encompass_Preferred
Select strength and cut protection with durable coated gloves Made with sturdy 10 gauge HPPE and steel shell Soft black nitrile foam coating Provides a solid grip both wet and dry Dark coloring hides dirt and grime CE Cut score 5 ANSI Cut Resistance- A5 ANSI Abrasion Resistance- Level 5 ANSI Puncture Resistance- Level 4 Anti-Vibration- No
Catalog No. 50-000-22342