Learn More
Essendant GLOVESKEVLARSPRYL
Supplier:  Essendant CRW9396S
Encompass_Preferred
Kevlar string knit gloves are used for general purpose work applications to provide cut and abrasion resistance using high performance yarns to further protect the hands when working with sharp objects The 9396 features a light weight 100pct DuPont Kevlar material with PVC dots on both sides ANSI Cut Resistance- A2 Anti-Vibration- No Size Type- Small Color(s)- Yellow/Black
Catalog No. 50-000-22314