Learn More
Essendant GLOVESLTX 92743LTMOR
Supplier:  Essendant CRW92743LTM
Encompass_Preferred
The 92743LT Cut Pro gloves feature our seamless orange HyperMax shell for long-lasting protection and are coated on the palm and over the knuckle with crinkle latex This glove provides outstanding cut protection and the crinkle latex coating provides superior gripping power The black coating hides dirt and grime making it look new and clean longer ANSI Cut Resistance- A8 ANSI Abrasion Resistance- Level 4 ANSI Puncture Resistance- Level 4 Anti-Vibration- No
Catalog No. 50-000-19461