Learn More
Essendant GLOVESNINJA FLEXMED
Supplier:  Essendant CRWN9680M
Encompass_Preferred
Sturdy gloves are designed with a crinkle latex coating for superb flexibility elasticity and durability Gloves provide excellent dexterity and protection Component materials comply with all relevant federal regulations for food contact ANSI Abrasion Resistance- Level 3 ANSI Puncture Resistance- Level 2 Anti-Vibration- No EN 388 Badge- 3131
Catalog No. 50-000-19127