Learn More
Essendant GLOVESNITRILE XLGEGY
Supplier:  Essendant ANS50709
Encompass_Preferred
A low cost nitrile glove combining the sensitivity of latex with the protection and strength of nitrile The thin mil gray gloves are ambidextrous powder-free with beaded cuffs and textured fingertips They are not made with natural rubber latex Glove Material- Nitrile Packaged Sterile- No Size Type- X-Large Length- 9 5
Catalog No. 50-000-20407