Learn More
Essendant GLOVESNITRILE4MM100
Supplier:  Essendant ANS54187CT
Encompass_Preferred
When you re looking for a glove that is designed for superior strength and tactile sensitivity KleenGuard G10 Comfort Plus Nitrile Gloves are an excellent choice These ambidextrous gloves feature a beaded cuff for easy donning textured fingertips for dexterity and grip and 4 mil thickness for comfort and ease of movement They are powder-free touchscreen compatible and are not made with natural rubber latex Glove Material- Nitrile Packaged Sterile- No Size Type- Medium Length- 9 5
Catalog No. 50-000-20350