Learn More
Essendant GLOVESNITRILE6MILXLBE
Supplier:  Essendant ANS54424
Encompass_Preferred
Thanks to the smart design these powder-free ambidextrous touchscreen compatible blue nitrile feature a 6 mil thickness for both comfort and protection The textured fingertips give employees a grippable surface for dexterity while performing sensitive tasks The beaded cuff helps prevent roll-down and makes the gloves easier to don these gloves are available in a variety of sizes are 9 5 long and are not made with natural rubber latex These blue nitrile gloves are a leading choice across a variety of industries because they are comfortable to wear provide excellent tactile sensitivity and deliver reliable protection for the task at hand KleenGuard G10 2Pro Nitrile Gloves meet US FDA regulations for food contact applications In addition these gloves are recyclable through The RightCycle Program by Kimberly-Clark Professional Making the right Personal Protection choice is difficult Perform with distinction with KleenGuard Personal Protective Equipment (PPE) Glove Material-
Catalog No. 50-000-20415