Learn More
Essendant GLOVESNITRILELG10WH
Supplier:  Essendant ANS62993
Encompass_Preferred
For critical (Class 3 and higher) cleanroom applications KIMTECH PURE G3 has you covered Close second-skin fit helps you maintain maximum dexterity Textured palm and fingers enable grip-sure performance Double-bag packaged in a Class 100 cleanroom AQL 1 5 rating Latex-free
Catalog No. 50-000-20588