Learn More
Essendant GLOVESNITRILEMD10WH
Supplier:  Essendant ANS62992
Encompass_Preferred
For critical (Class 3 and higher) cleanroom applications KIMTECH PURE G3 has you covered Close second-skin fit helps you maintain maximum dexterity Textured palm and fingers enable grip-sure performance Double-bag packaged in a Class 100 cleanroom AQL 1 5 rating Latex-free Glove Material- Nitrile Packaged Sterile- No Size Type- Medium Length- 12
Catalog No. 50-000-20908